Find methods for your needs
Fast analysis of uron herbicides using a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of uron herbicides. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column, with UV detection at 240 nm.| Market: | Environmental |
| Keywords: | Accucore RP-MS, Chlortoluron, Diuron, HPLC, Linuron, Metoxuron, Monuron, Terbuthiuron, Uron herbicides, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.