Find methods for your needs
Improved analysis of isopropylthioxanthones using a Thermo Scientific Hypersil GOLD PFP HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of isopropylthioxanthones. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 262 nm.| Market: | Chemical |
| Keywords: | HPLC, Hypersil Gold PFP, Isopropylthioxanthones, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.