Find methods for your needs
Improved analysis of phenolic positional isomers using a Thermo Scientific Hypersil GOLD PFP HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of phenolic positional isomers. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 270 nm.| Market: | Chemical |
| Keywords: | HPLC, Hypersil Gold PFP, Phenolic positional isomers, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.