Find methods for your needs
Improved analysis of phenolic positional isomers using a Thermo Scientific Hypersil GOLD PFP HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of phenolic positional isomers. The separation is performed on a Thermo Scientific Hypersil GOLD PFP HPLC column, with UV detection at 270 nm.
Market: Chemical
Keywords: HPLC, Hypersil Gold PFP, Phenolic positional isomers, UV detection
Matrix: N/A
Uploaded on 9/3/2012.
For Research Use Only. Not for use in diagnostic procedures.