Find methods for your needs
Analyzing PBDEs in House Dust Samples with the Thermo Scientific TSQ Quantum XLS Ultra GC-MS/MS in EI-SRM Mode
Description
Polybrominated diphenyl ethers (PBDEs) were introduced in the 1960s as flame retardants and are currently used in a wide variety of household apparatuses, consumer electronics, furniture and more. Environmental levels of PBDEs have been continuously increasing due to their special persistence.Due to growing concerns over the health risks from constant exposure to this class of compounds and the accumulation effects in the food chain, suitable analytical methods are required to quantify flame retardant compounds at low levels in a variety of matrices. The new range of Trace Gold Metal columns aim to increase the lifetime of the GC column due to the more robust nature of the GC column. Chromatographic performance is not compromised when selecting a TraceGOLD TG-5MT column over the equivalent TG-5MS column.| Market: | Environmental |
| Matrix: | House dust sample |
For Research Use Only. Not for use in diagnostic procedures.