Find methods for your needs
Determination of inorganic disinfection byproducts, bromide, and common inorganic anions in packaged drinking water and tap water using ion chromatography Be the first to rate this application
dbp anions analysis from water samples
Description
This application note describes a reagent-free ion chromatography (RFIC) method for determining trace oxyhalide disinfection byproducts (chlorite, bromate, and chlorate), bromide, and common inorganic anions in packaged drinking water and tap water. Using the Dionex IonPac AS19-4μm column, NGES-A suppressor, KOH eluent, and suppressed conductivity detection, the method achieves 0.50 μg/L detection with <2% RSD and 97–102% recoveries.
Market: Environmental
Keywords: Bromate, Chlorate, Chlorite, environmental, fluoride, Nitrate, RFIC, Nitrite, Chloride, phosphate, Sulfate, IonPac AS19-4μm, NGES-A
Matrix: Packaged drinking water, Tap water
Author: Chanakya Thaker; Neil Rumachik
Affiliation: Thermo Fisher Scientific
Uploaded on 9/24/2026.
For Research Use Only. Not for use in diagnostic procedures.