Find methods for your needs
AN004644 Determination of total fluorine, chlorine, and sulfur in liquefiedpetroleum gas using combustion-ion chromatography Be the first to rate this application
an004644 halogens sulfur lpg
Description
Natural gas, an important energy source, is often compressed into liquid for easier transportation. Combustion-IC (C-IC) has been previously demonstrated as an ideal approach to eliminate the sample matrix and increase sample homogeneity for various sample matrices. Additionally, C-IC is recognized as an ASTM standardized method (D7994) to determine halogens and sulfur in LPG. Here, we determine halides and sulfur species in LPG using C-IC, leveraging advances in the Thermo Scientific™ Cindion™ C-IC system using the Thermo Scientific™ Cindion™ LPG/Gas Module with built-in safety features.
Market: Petrochem, Oil and Gas
Keywords: fluoride, Chloride, Sulfur, Bromide, LPG, combustion IC, Inuvion, Cindion C-IC, combustion-IC, IonPac AS18 Fast, Liquefied Natural Gas, Liquefied Petroleum Gas, LNG
Matrix: butane LPG, Liquefied Natural Gas, Liquefied Petroleum Gas, LNG, LPG
Author: Terri Christison and Neil Rumachik
Affiliation: Thermo Fisher Scientific
Uploaded on 7/2/2026.
For Research Use Only. Not for use in diagnostic procedures.