Find methods for your needs
USP-aligned ion chromatographic assay of magnesium sulfate using a next-generation electrolytic suppressor Be the first to rate this application
magnesium sulfate assay using ic
Description
This application note demonstrates a USP-aligned ion chromatography method for quantitative assay of magnesium sulfate using the Dionex IonPac CS16 column and NGES-C electrolytic suppressor. The method uses suppressed conductivity detection with 48 mM MSA and meets USP system suitability criteria, including Mg/Ca resolution, tailing factor, and %RSD. Linearity was confirmed from 10–300 µg/mL, and sample assays fell within 98–102% with low RSDs. The NGES-C improves baseline stability, noise reduction, and robustness for routine pharmaceutical QC.
Market: Pharma; BioPharma
Keywords: Pharmacopeia, USP, IonPac CS16 column, NGES-C, Pharmaceuticals: NGES suppressor
Matrix: Magnesium Sulfate
Author: Chanakya Thaker; Neil Rumachik
Uploaded on 5/27/2026.
For Research Use Only. Not for use in diagnostic procedures.