Find methods for your needs
Determination of nitrite and nitrate from lactose by ion chromatography using the NGES-A suppressor Be the first to rate this application
nitrite nitrate analysis from lactose using ic
Description
This application note presents an ion chromatography method for trace nitrite and nitrate analysis in lactose monohydrate and colloidal silicon dioxide. Using a Dionex IonPac AS19-4µm column and NGES-A suppressor, the method achieved sensitive, selective separation with LOQs of 0.1 µg/g nitrite and 0.2 µg/g nitrate. Strong linearity, precision, and recovery support its use in USP-aligned nitrosamine risk assessment for pharmaceutical excipients
Market: Pharma; BioPharma
Keywords: Excipients, Pharmaceuticals, USP, IonPac AS19 column, NGES, suppressor
Matrix: Colloidal Silicon Dioxide, Lactose
Author: Chanakya Thaker; Neil Rumachik
Affiliation: Thermo Fisher Scientific
Uploaded on 5/27/2026.
For Research Use Only. Not for use in diagnostic procedures.