Find methods for your needs
AN268: AN268, additional data: Effect of dissolved iron in MSA eluent when determining chelators (5 mg/L EDTA, NTA, DTPA, EGTA). Be the first to rate this application
an268 effect dissolved iron eluent
Description
This is additional data not reported in AN268. Shows the effect of a Dionex IonPac CTC-1 trap column trapping interfering dissolved iron from manually prepared MSA eluent. This trap can be used in other applications where acidic eluents might have dissolved metal contamination.
Market: Environmental
Keywords: EDTA, EGTA, NTA, DTPA, Pt working electrode, IonPac AS7 column, PAD, ED detection, chelator, CTC-1 trap column, dissolved iron, Pt waveform
Matrix: Drinking water, gray water, Wastewater
Author: Terri Christison, Brian De Borba, and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 5/1/2026.
For Research Use Only. Not for use in diagnostic procedures.