Find methods for your needs
Quantitation of an oral fluid drug panel including THC using the Stellar mass spectrometer
Description
The Thermo Scientific™ Vanquish™ Horizon UHPLC coupled with the Thermo Scientific™ Stellar™ mass spectrometer is used for the separation and detection of drugs of abuse in oral fluid matrix. The SPE extraction uses XTR tips with mixed mode SCX/WAX chemistry. Using a Thermo Scientific™ Accucore™ Biphenyl column with a total run time of 4.5 minutes, all isomers were separated, including methamphetamine / phentermine and hydromorphone / morphine. The method includes both CID (resonance-type, unique to ion traps) and HCD (beam-type) activations for more optimized fragmentation.| Market: | Clinical Research |
| Keywords: | drugs of abuse, Toxicology, clinical, Forensics, Oral Fluid |
| Matrix: | oral fluid |
For Research Use Only. Not for use in diagnostic procedures.