Find methods for your needs
Enhanced analytical flexibility: simultaneous normal and reversed phase chromatography Be the first to rate this application
Description
Use Vanquish Flex Duo UHPLC with NP kit to run NP (vitamin D3) and RP (B1, B2, B3, B6) simultaneously for USP-aligned, high-accuracy vitamin analysis in pregnancy capsules.
Market: Food and Beverage
Keywords: Cholecalciferol, Niacinamide, Normal phase, Riboflavin, Thiamine, Water-soluble vitamins, Fat-soluble vitamins, Pyridoxine, Vitamin D, Reversed phase, Vanquish Flex Duo UHPLC System, pregnancy and breastfeeding vitamins, Vitamin B
Matrix: n-hexane
Author: Sandra Kmieliauskaitė, Sylvia Grosse, Jan Moebius
Affiliation: Thermo Fisher Scientific, Vilnius; Thermo Fisher Scientific, Germering, Germany
Uploaded on 3/5/2026.
For Research Use Only. Not for use in diagnostic procedures.