Find methods for your needs
Enhanced analytical flexibility: simultaneous normal and reversed phase chromatography
Description
Use Vanquish Flex Duo UHPLC with NP kit to run NP (vitamin D3) and RP (B1, B2, B3, B6) simultaneously for USP-aligned, high-accuracy vitamin analysis in pregnancy capsules.| Market: | Food and Beverage |
| Keywords: | Cholecalciferol, Niacinamide, Normal phase, Riboflavin, Thiamine, Water-soluble vitamins, Fat-soluble vitamins, Pyridoxine, Vitamin D, Reversed phase, Vanquish Flex Duo UHPLC System, pregnancy and breastfeeding vitamins, Vitamin B |
| Matrix: | n-hexane |
| Author: | Sandra Kmieliauskaitė, Sylvia Grosse, Jan Moebius |
| Affiliation: | Thermo Fisher Scientific, Vilnius; Thermo Fisher Scientific, Germering, Germany |
For Research Use Only. Not for use in diagnostic procedures.