Find methods for your needs
Robust peptide quantitation using the TSQ Certis triple quadrupole mass spectrometer and OptiSpray ion source and cartridges Be the first to rate this application
robust peptide quantitation using tsq certis triple quadrupole mass spectrometer optispray ion source cartridges
Description
This work demonstrated quantitative targeted peptide analysis using the TSQ Certis triple quadrupole mass spectrometer coupled with the Vanquish Neo UHPLC system. With the OptiSpray ion source and cartridge columns optimized for nano- and capillary flow (<5 µL/min), the platform delivered strong reproducibility, retention time stability, and sensitivity for an 82-peptide plasma biomarker panel.
Market: Proteomics
Keywords: Vanquish Neo UHPLC, OptiSpray ion source and cartridge, TSQ Certis mass spectrometer
Author: Qin Fu et al.
Affiliation: Thermo Fisher Scientific
Uploaded on 2/17/2026.
For Research Use Only. Not for use in diagnostic procedures.