Find methods for your needs
Online SPE-UHPLC workflow for the determination of bitter acids in hops Be the first to rate this application
Description
α- and β-bitter acids in hops were determined using the Thermo Scientific™ Vanquish™ Online SPE UHPLC system. Online SPE with a Hypersil GOLD™ C8 column (20 × 2.1 mm) eliminates the need for standalone SPE cartridges, while separation of α- and β-acids and their derivatives is achieved using a Hypersil GOLD™ C18 Selectivity column.
Market: Food and Beverage; Environmental
Keywords: Humulones, Hypersil GOLD, Online SPE, lupulones
Matrix: hops
Author: Elīna Pasečnaja
Affiliation: Thermo Fisher Scientific, Vilnius, Lithuania
Uploaded on 1/30/2026.
For Research Use Only. Not for use in diagnostic procedures.