Find methods for your needs
AN 003644: Comprehensive screening of per- and polyfluoroalkyl substances (PFAS) in food contact materials
Description
We developed a sensitive and automated method using CIC to determine both TOF and EOF in FCM. The TOF method is beneficial for manufacturers to comply with current state regulations. We also employed a non-targeted LC-HRMS approach, which allowed us to detect 46 PFAS compounds in the EOF fraction, many of which fall outside of the typical list of targeted screening or quantification methods.| Market: | Food and Beverage |
| Keywords: | Combustion ion chromatography (CIC), extractable organic fluorine (EOF), total organic fluorine (TOF), food contact materials (FCM), non-targeted analysis (NTA) |
| Matrix: | food contact material |
| Author: | Jingli Hu, Richard Cochran, Cynthia Grim, Neil Rumachik |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.