Find methods for your needs
Monitoring monoclonal antibody bioproduction processes with 2D-LC-UV Be the first to rate this application
Description
The switchable heart-cut 2D-LC system facilitates the straightforward at-line analysis of mAb producing cell culture samples for the upstream monitoring of titer, charge variants, and size variants of the intact antibody. While the first dimension purifies the mAb from matrix components by Protein A affinity chromatography, selectable second dimension mechanisms enable the assessment of the aggregation level by SEC or the abundance monitoring of charge variants by SCX.
Market: BioPharma
Keywords: Size exclusion, SEC, mAb, Cation Exchange, Protein A, SCX, Charge Variants, Process control, heart-cut 2D-LC, size variants
Matrix: Cell culture, HCC
Author: Maria Grübner
Affiliation: Thermo Fisher Scientific, Germering, Germany
Uploaded on 12/1/2025.
For Research Use Only. Not for use in diagnostic procedures.