Find methods for your needs
AB004042 IC-MS analysis of perchlorate with a new, robust, and reliable next-generation suppressor Be the first to rate this application
Description
Demonstrating the performance of the Thermo Scientific Dionex NGES Anion Suppressor (NGES-A) for the implementation of US EPA Method 332.0, highlighting the increased robustness and resilience compared to the previous suppressor model.Design enhancements of the new Dionex NGES suppressor ensure reliable analyses to meet the growing demands of the IC hyphenated solutions such as IC-MS.EPA 332.0
Market: Environmental
Keywords: IC-MS, EPA 332, electrolytic suppressor, NGES
Matrix: Drinking water
Author: Yukiko Kawahara, Neil Rumachik
Uploaded on 11/25/2025.
For Research Use Only. Not for use in diagnostic procedures.