Find methods for your needs
Comprehensive profiling of plastic polymers using pyrolysis coupled to GC-MS
Description
The aim of this study is to demonstrate the suitability of pyrolysis coupled to GC-MS (Py-GC-MS) for the identification and quantitation of the 12 most common polymer types within a single GC-MS run as well as qualitative analysis of plastic samples collected on the beach.| Market: | Environmental |
| Keywords: | Gas Chromatography, single quadrupole mass spectrometry, pyrolysis, TRACE 1610 GC, ISQ 7610 GC-MS, Plastic polymers, microplastics (MPs), Py-GC-MS |
| Matrix: | Sand |
| Author: | Giulia Riccardino[[1]], Manuela Bergna[[1]], Daniela Cavagnino[[1]], and Daniel Kutscher [[2]] |
| Affiliation: | [[1]] Thermo Fisher Scientific, Milan, IT [[2]] Thermo Fisher Scientific, Bremen, UK |