Find methods for your needs
Amino acid analysis in food, beverages, and fertilizers by automated in-needle OPA/FMOC derivatization Be the first to rate this application
Description
A time and labor-saving online sample preparation workflow by amino acid derivatization in the autosampler's needle is presented for medium and highly complex sample matrices (beverages, food, and fertilizers) analyzed at one external and two in-house laboratories. Robust and reproducible method performance, column stability over several hundred injections, and applicability at different instrument tiers are highlighted.
Market: Food and Beverage
Keywords: Amino acids, Derivatization, UDP, Vanquish Flex UHPLC, Accucore column, custom injection program, automation, Vanquish Core HPLC, FMOC, OPA, CIP
Matrix: Beer, fertilizer, Honey, Juice, soy sauce, Wine
Author: Maria Grübner, Andreas Becker, Pau Castells
Affiliation: Thermo Fisher Scientific
Uploaded on 4/8/2025.