Find methods for your needs
AN003548 Fast analyses of anions in water with microbore columns using a compact ion chromatography system for reduced eluent use
Description
Industries are adopting methods to enhance analytical performance while minimizing ecological impact. In ion chromatography, using microbore columns with diameters of 2 mm or less reduces eluent use, cutting hazardous waste and improving throughput.This note highlights rapid anion analysis in water using microbore column, which significantly reduces liquid waste compared to standard bore columns. This method enhances safety and sustainability by minimizing waste production.Both chemical and electrolytic suppression techniques are demonstrated.| Market: | Environmental |
| Keywords: | Chemical Suppression, EPA 300.1, electrolytic suppression |
| Matrix: | Drinking water, Surface water, Waste water |
| Author: | Yukiko Kawahara, Neil Rumachik |
For Research Use Only. Not for use in diagnostic procedures.