Find methods for your needs
Sensitive and cryogen-free analysis of epichlorohydrin and other VOCs in drinking water by using In-Tube Extraction Dynamic Headspace (ITEX-DHS) sampling coupled to GC-MS
Description
The aim of this study is to demonstrate the suitability of In-Tube Extraction Dynamic Headspace (ITEX-DHS) for the analysis of very volatile organic compounds (VVOCs), volatile organic compounds (VOCs), and epichlorohydrin (ECH) in drinking water, when coupled to a cryogen-free refocusing into a programmed temperature vaporizer (PTV) injector.| Market: | Environmental |
| Keywords: | Drinking Water, environmental, Gas Chromatography, single quadrupole mass spectrometry, VOCs, TRACE 1610 GC, ISQ 7610 GC-MS, Epichlorohydrin, dynamic headspace, ECH, ITEX-DHS, volatile organic compounds, VVOCs |
| Matrix: | Drinking water |
| Author: | Giulia Riccardino[[1]] , Manuela Bergna[[1]] , Daniela Cavagnino[[1]] , and Daniel Kutscher[[2]] |
| Affiliation: | [[1]] Thermo Fisher Scientific, Milan, IT [[2]] Thermo Fisher Scientific, Bremen, DE |
For Research Use Only. Not for use in diagnostic procedures.