Find methods for your needs
Fast analysis of ondansetron using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of ondansetron. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Antiemetic, Anti-nausea, HPLC, Hypersil GOLD, Ondansetron, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.