Find methods for your needs
AN003262: Analysis of common lithium salts, trace additives, and contaminants in lithium-ion battery electrolytes by ion chromatography-mass spectrometry
Description
An ion chromatography-mass spectrometry (IC-MS) method to meet the analytical need of understanding the sample composition and degradation byproduct in simulated battery electrolytes. Integrating an organic solvent gradient, this method achieves excellent separation of many analytes allowing lithium-ion battery (LIB) manufacturers and researchers to gain knoledge to improve quality assurance, quality control and failure analysis processes to ensure the battery's performance.| Market: | Chemical |
| Keywords: | IC-MS, ISQ EC, CRD 300 carbonate removal device, Dionex IonPac AS23 |
| Matrix: | Battery Electrolyte |
| Author: | Yukiko Kawahara, Detlef Jensen, Neil Rumachik |