Find methods for your needs
AN002748: EPA Method 1621 collaboration study_Combustion IC
Description
Per- and polyfluoroalkyl substances (PFAS) is the collective name for synthetic fluorinated compounds. Here we describe our lab's results from the EPA draft Method 1621 multi-lab study results to screen for PFAS using adsorbable organic fluorine by combustion ion chromatography. The recovery of 4 types of PFAS from wastewater samplers are discussed. This method was found to be robust, precise, and accurate. Additionally, a technical note is included to describe tips and tricks for successful implementation of EPA Method 1621.| Market: | Environmental |
| Keywords: | waste water, fluoride, IonPac AS24 column, PFOA, PFOS, CIC, wastewater, Integrion, PFAS, combustion IC, AOX, polyfluoroalkyl substances, AOF, PFBA, pyrohydrolytic combustion IC, perfluorinated, Perfluoroalkyl substances, GAC, collaboration study, EPA Method 1621, Dionex IonPac AS24 column, AS24 column, multi-lab study, tn 003056, successful implementation of EPA 1621, techniques successful EPA 1621, tn003056 |
| Matrix: | Wastewater, Water |
| Author: | Terri Christison and Neil Rumachik |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.