Find methods for your needs
AN73883 Determination of per- and polyfluorinated alkyl substances (PFAS) in drinking water Using automated solid phase extraction and LC-MS/MS for U.S. EPA Method 533
Description
Drinking water perfluorinated and polyfluorinated alkyl substances (PFAS) studies have typically targeted PFOS and PFBA and other PFAS. Here we discuss the development of an analytical method using an automated SPE system and LC-MS/MS for determination of twenty-five PFAS compounds following the guidelines provided by U.S. EPA Method 533. We have demonstrated that the automated SPE system provides reliable determination of PFAS in large-volume aqueous samples and saves time, solvent, and labor, while ensuring high reproducibility and productivity for analytical testing laboratories.| Market: | Environmental |
| Keywords: | Acclaim 120 C18 Column, PFOA, PFOS, Vanquish Flex UHPLC, perfluorinated alkyl substances, PFAS, PFHxS, PFNA, PFHpA, PFBA, PFHxA, AutoTrace 280 PFAS, polyfluorinated alkyl substances, U.S. EPA Method 533, TSQ Fortis triple quadrupole mass spectrometer, PFUnDA |
| Matrix: | Drinking water |
| Author: | Xin Zhang, Changling Qiu, Rahmat Ullah, and Yan Liu |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.