Find methods for your needs
AN002771 LC-MS/MS analysis of per- and polyfluoroalkyl substances (PFAS) in soil samples in accordance with EPA Method 1633: semi-automated solid phase extraction cleanup
Description
PFAS workflows are continuously being developed to meet evolving testing requirements for per- and polyfluoroalkyl substances (PFAS) worldwide. This work describes that workflow by leveraging the Dionex AutoTrace 280 PFAS SPE instrument to automate the soil sample solid-phase extraction (SPE) cleanup stage that is described in EPA Method 1633.| Market: | Environmental |
| Keywords: | Acclaim, Hypersil GOLD, SPE, PFOA, PFOS, AutoTrace 280, PFAS, EPA Method 1633, PFHpS, PFHxS, PFTrDA, PFDA, PFNA, PFHpA, PFDoA, AN 002771, AN00771, HFPOA-DA, N-EtFOSA, N-EtFOSE, NFDHA, N-MeFOSA, N-MeFOSE, PFBA, PFBS, PFDoS, PFDS, PFEESA, PFHxA, PFMBA, PFMPA, PFNS, PFOSA, PFPeA, PFPeS, PFTeDA, PFUdA, semi-automated solid phase extraction |
| Matrix: | Soil samples |
| Author: | Cynthia Grim1, Kevin McHale2, and Toby Astill1 |
| Affiliation: | 1Thermo Fisher Scientific, San Jose, CA. 2Thermo Fisher Scientific, Somerset, NJ |
For Research Use Only. Not for use in diagnostic procedures.