Find methods for your needs
Chiral analysis of Vilanterol Trifenatate using a Hypersil Chiral IGT column
Description
The Thermo Scientific UltiMate 3000 RS HPLC System is applied for the chiral purity of (S)-Isomer in Vilanterol Trifenatate. The analysis was performed on a Thermo Scientific Hypersil Chiral IGT HPLC column using an in-house method. The result obtained for resolution between S-isomer and Vilanterol Trifenatate is 4.12.The results obtained for system suitability were 2.37 for the S-isomer and Vilanterol Trifenatate were 1.0 and 1.02 respectively.Market: | Pharma |
Matrix: | Diluent |
Author: | Mayuresh Chiplunkar |
Affiliation: | ThermoFisher Scientific CSC-Mumbai |