Find methods for your needs
Charg varient Analysis of Tocilizumab Be the first to rate this application
Description
The Thermofisher Scientific Vanquish™ Flex UHPLC system was used to analyze Tocilizumab Charge variant analysis using Propac 3R SCX Column and CX-1 Buffer
Market: BioPharma
Keywords: Vanquish Flex, Propac 3R SCX, CX-1 buffer, Tocilizumab Charge varient
Matrix: Formulation
Author: Mazin Adenwala
Affiliation: Thermo Fisher Scientific, CoE Lab, Ahmedabad, India
Uploaded on 11/29/2023.