Find methods for your needs
Charge Varient analysis of Obinutuzumab on Propac 3R SCX Be the first to rate this application
Description
The Thermofisher Scientific Vanquish™ Flex UHPLC system was applied for the analysis of Charge related impurities in obinutuzumab using Propac 3R Column and CX-1 buffer
Market: BioPharma
Keywords: Vanquish Flex, Propac 3R SCX, CX-1 buffer, Obinutuzumab IEX
Matrix: Formaulation
Author: Mazin Adenwala
Affiliation: Thermo Fisher Scientific, CoE Lab, Ahmedabad, India
Uploaded on 11/29/2023.