Find methods for your needs
Impurity Analysis of Caspofungin using Acclaim C30 HPLC Column Be the first to rate this application
Description
The Thermo Scientific™ Vanquish™ Flex UHPLC system is applied for the Related substance analysis of Caspofungin. The analysis was performed on the Thermofisher Scientific Acclaim C30 HPLC Column. The results obtained for the Resolution and theoretical plates met the criteria.
Market: Pharma
Keywords: Acclaim C30, Vanquish Flex, Caspofungin RS method
Matrix: NA
Author: Mazin Adenwala
Uploaded on 11/21/2023.
For Research Use Only. Not for use in diagnostic procedures.