Find methods for your needs
APN002340: Fast determination of anions in municipal drinking water using a compact ion chromatography system with a chemically regenerated suppressor
(1)
Description
Demonstrates the determination of inorganic anions in drinking water by IC with built-in peristaltic pump for regenerant flow to a chemical suppressor.
Market: Environmental
Keywords: suppressed conductivity detection, U.S. EPA Method 300.1, Clean Water Act(CWA), Safe Drinking Water Act(SDWA)
Matrix: Drinking water
Uploaded on 11/15/2023.