Find methods for your needs
APN 002342 Determination of trace concentrations of oxyhalides and bromide in municipal and bottled waters
Description
The method used a Thermo Scientific™ Dionex™ IonPac™ AS23-4μm column, a Thermo Scientific™ Dionex™ AERS™ 500 Carbonate anion electrolytically regenerated suppressor, and a Thermo Scientific™ Dionex™ CRD 300 Carbonate Removal Device in vacuum mode. This work demonstrates a similar method on a Thermo Scientific™ Dionex™ Inuvion™ ion chromatography system, a new integrated, single-channel compact IC system.| Market: | Environmental |
| Keywords: | Bromate, RFIC, suppressed conductivity detection, Dionex IonPac AS23-4μm column, DBPs, U.S. EPA Method 300.1, Safe Drinking Water Act (SDWA), Clean Water Act (CWA), Dionex Inuvion system, Inorganic disinfection by-products, peristaltic pump, EGC K2CO3 cartridge, P/N 088453 |
| Matrix: | Water |
| Author: | Hua Yang, Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.