Find methods for your needs
APN 002311: Assay for citrate and phosphate in pharmaceutical formulations using a compact IC system
Description
This application proof note demonstrates a determination of citrate and phosphate in pharmaceutical formulations using the method published in Thermo Fisher Scientific Application Note 164. In this proof note, the method is performed using Inuvion IC system.| Market: | Pharma |
| Keywords: | USP, Dionex IonPac AS11 column, Dionex ADRS 600 suppressor, Dionex Inuvion IC system |
| Matrix: | Water |
| Author: | Jingli Hu, Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.