Find methods for your needs
APN002384: Determination of ammonia in sodium bicarbonate using a compact RFIC ion chromatography system
Description
Sodium bicarbonate, or sodium hydrogen carbonate, is used in a wide variety of pharmaceutical and food products, and ammonia is one of the impurities that is assayed. In this application proof note, the performance of the easy-to-use, compact Thermo Scientific Dionex Inuvion ion chromatography system was evaluated for this application.Market: | Pharma; Food and Beverage |
Keywords: | USP, Dionex IonPac CS16 Column, Sodium, Ammonium, Dionex Inuvion IC system, bicarbonate |
Matrix: | Sodium Bicarbonate |
Author: | Yukiko Kawahara, Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |