Find methods for your needs
AN 002344 : Determination of fluoride in sodium fluoride oral solution using carbonate/bicarbonate eluent
Description
This application developed a new method for the determination of fluoride in oral rinse products using carbonate/bicarbonate eluent. The method demonstrates excellent accuracy and precision for fluoride in commercially available oral rinse samples and, therefore, can be used as a quality control method by oral rinse manufacturers.| Market: | Pharma |
| Keywords: | Mouthwash, ion chromatography, Dental, Dionex ICS-6000 HPIC system, Dionex IonPac AS22 column, Dionex ADRS 600 suppressor, mouth rinse |
| Matrix: | Water |
| Author: | Jingli Hu, Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.