Find methods for your needs
Ropinorile assay as per USPNF 2021 ISSUE 1 on UltiMate 3000 HPLC system Be the first to rate this application
Description
The Thermo Scientific UltiMate 3000 RS HPLC System is applied for the assay of Ropinirole. The analysis was performed on a Thermo Scientific Hypersil Gold C8 HPLC column using USPNF 2021 ISSUE 1 method. The results obtained for % RSD of Standard solution meets the criteria stated in USPNF 2021 ISSUE 1. The results obtained for standard solution is 0.4 % USP criteria NMT 1.0 %. The tailing factor criteria stated NMT 1.6 and obtained results was 1.07 on UltiMate 3000 RS HPLC System.
Market: Pharma
Matrix: Diluent
Author: Mayuresh Chiplunkar
Affiliation: ThermoFisher Scientific CSC-Mumbai
Uploaded on 8/31/2023.