Find methods for your needs
GRANISETRON HCL ASSAY-USP43-NF38 Be the first to rate this application
Description
GRANISETRON HCL ASSAY-USP43-NF38 using a Thermo Scientific UltiMate 3000 HPLC system. The resolution observed was 12.36, while the limit is NLT: 3.5. The tailing factor observed was 1.20. The limit is NMT 2.0 for system suitability solution. % RSD for standard solution observed was 0.13 %. The limit NMT 0.73 %.
Market: Pharma
Matrix: Diluent as mobile phase
Author: Mayuresh Chiplunkar
Affiliation: ThermoFisher Scientific CSC-Mumbai
Uploaded on 8/22/2023.