Description
GRANISETRON HCL ASSAY-USP43-NF38 using a Thermo Scientific UltiMate 3000 HPLC system. The resolution observed was 12.36, while the limit is NLT: 3.5. The tailing factor observed was 1.20. The limit is NMT 2.0 for system suitability solution. % RSD for standard solution observed was 0.13 %. The limit NMT 0.73 %.| Market: | Pharma |
| Matrix: | Diluent as mobile phase |
| Author: | Mayuresh Chiplunkar |
| Affiliation: | ThermoFisher Scientific CSC-Mumbai |