Find methods for your needs
Rivaroxaban enantiomeric purity as per USPNF 2022 on UltiMate 3000 RS HPLC System.
Description
The enantiomeric purity of Rivaroxaban was determined on a Thermo Scientific UltiMate 3000 RS HPLC System. The analysis was performed on a Thermo Scientific Hypersil Chiral ICT column using the USPNF 2022 method. The results obtained for % RSD of standard solution meets the criteria stated in USPNF 2022. The results obtained for standard solution is 0.87 % USP criteria NMT 5.0 %. The resolution criteria stated NMT 1.5 and obtained results was 3.18 on UltiMate 3000 RS HPLC system.| Market: | Pharma |
| Matrix: | Diluent |
| Author: | Mayuresh Chiplunkar |
| Affiliation: | ThermoFisher Scientific CSC-Mumbai |