Find methods for your needs
APN002416 Sensitive determinations of hexavalent chromium in drinking water using a compact IC system Be the first to rate this application
Description
Chromate and dichromate are colloquially known as hexavalent chromium, Cr(VI). Hexavalent chromium is potentially a carcinogenic agent; therefore, it is a regulated compound. Total chromium as chromate is determined according to U.S. EPA Method 218.6 or slightly modified U.S. EPA Method 218.7 in Application Update 179. In this application proof note, the same application is demonstrated on a 2 mm i.d., Dionex IonPac AS7 anion-exchange column. Chromate is detected at 530 nm, after adding a post-column reagent. The application is facilitated by the Thermo Scientific™ Dionex™ Inuvion™ IC system.
Market: Environmental
Keywords: PCR, Cr(VI), hexavalent chromium, chromate, EPA 218.6, Dionex Inuvion IC system, AU179, EPA 218.7
Matrix: High Ionic Water, Municipal drinking water
Author: Terri Christison and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 8/9/2023.
For Research Use Only. Not for use in diagnostic procedures.