Find methods for your needs
AN002343: Determination of Chloride and Sulfate in Saturated Lithium Hydroxide Solution
Description
This application note describes a method for the determination of chloride and sulfate in saturated lithium hydroxide solution using an RFIC system with a 2 mm version of a Dionex IonPac AS29-Fast-4μm column. The method is sensitive (MDL = 0.09 mg/L for chloride and 0.13 mg/L for sulfate in saturated lithium hydroxide solution), precise (RSD range = 1–6%), and accurate (recovery range = 91–102%).| Market: | Chemical; Energy; Material Science |
| Keywords: | suppressed conductivity detection, RFIC System, Dionex IonPac AS29-Fast-4μm column, lithium-ion batteries |
| Matrix: | saturated lithium hydroxide solution |
| Author: | Hua Yang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.