Find methods for your needs
Minocycline periodontal System assay and organic impurities as per USP43-NF38 on Thermo Scientific Vanquish UHPLC system.
Description
The Thermo Scientific Vanquish UHPLC system was used for the assay and organic Impurities of minocycline Periodontal System. The analysis was performed on a Thermo Scientific Accucore XL-C8 HPLC column using the method described in the USP43-NF38 . The results obtained for resolution and tailing factor met the criteria stated in the USP43-NF38. The result obtained for resolution between the peaks due to epiminocylcine and minocycline was 4.8 (USP criteria of minimum 2.0 ).| Market: | Pharma |
| Matrix: | NA |
| Author: | Mayuresh Chiplunkar |
| Affiliation: | ThermoFisher Scientific CSC-Mumbai |