Find methods for your needs
AN001967: Determination of Inorganic Anions in Saturated Lithium Carbonate Solution
Description
This application note describes a method for the determination of inorganic anions (fluoride, chloride, nitrite, bromide, nitrate, phosphate, and sulfate) in saturated lithium carbonate solution using a RFIC system with a Dionex IonPac AS23 column, carbonate eluent suppressor, and carbonate removal device. The method is sensitive (MDL 0.02–0.22 mg/L in saturated lithium carbonate solution), precise (RSD range 1–6%, and accurate (recovery range 95–107%).| Market: | Chemical | 
| Keywords: | suppressed conductivity detection, CRD 300, RFIC System, Dionex IonPac AS23 column, lithium-ion batteries, Carbonate Eluent Suppressor, AERS 500 Carbonate, AN 001967, EPM, K2CO3 | 
| Matrix: | Saturated lithium carbonate solution | 
| Author: | Hua Yang and Jeffrey Rohrer | 
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA | 
For Research Use Only. Not for use in diagnostic procedures.