Find methods for your needs
CN001941: Determination of Chlorate in Agar Samples using Ion Chromatography (IC)
Description
The European Food Safety Authority (EFSA) reported chlorate as a risk to human health. In June 2020, the European Commission set chlorate limits for various foods, including the limit for algae (0.05 mg/kg). This work describes a straightforward method to determine chlorate in seaweed-derived agar products. A sample preparation circumvents unwanted gelation, thus avoiding clogging capillaries and columns. Our approach is suitable for analyzing chlorate where the final agar content does not exceed 0.5–1.0%| Market: | Food and Beverage |
| Keywords: | Bromate, Chlorate, Chlorite, Formate, Citrate, IonPac AS19 column, fluoride, Nitrate, RFIC, Agar, Nitrite, Chloride, phosphate, Sulfate, Algae, Reagent-Free IC, Bromide, Reagent-Free IC (RFIC), EFSA, Gelidium, Gracilaria, red algae, algal, seaweed, CN 001941 |
| Matrix: | agar |
| Author: | Tiziana Ladisa1 , Flavio Zanetti2 , Massimiliano Borgogna2 , Lidia Graziano3 , Detlef Jensen4 |
| Affiliation: | 1 Thermo Fisher Scientific, Rodano MI, Italy 2 Biopolife Srl, Trieste, Italy 3 Java Biocolloid Europe Srl, Trieste, Italy 4 Thermo Fisher Scientific GmbH, Dreieich, Germany |
For Research Use Only. Not for use in diagnostic procedures.