Find methods for your needs
Managing the challenges of analyzing brine solutions of variable concentration using inductively coupled plasma mass spectrometry (ICP-MS) equipped with argon gas dilution Be the first to rate this application
brine analysis using an icpms
Description
To highlight how ICP-MS can be used to determine impurities in highly concentrated brine solution (up to 25% w/w) with high sensitivity, accuracy, and robustness using automated sample dilution with argon gas delivered from the instrument.
Market: Environmental
Keywords: High resolution, ICP-MS, KED, high matrix, single quadrupole, Argon gas dilution, brine solution
Matrix: Brine up to 25%, Sea water
Author: Tomoko Vincent, Daniel Kutsher
Affiliation: Thermo Fisher Scientific, germany
Uploaded on 4/14/2023.
For Research Use Only. Not for use in diagnostic procedures.