Find methods for your needs
TN001900: Evaluation of Cationic Quaternary Amines by Ion Chromatography and Mass Spectroscopy (IC-MS)
Description
Pesticide contamination in food as a potential health risk is a growing public concern. The determinations of cationic quaternary amine polar pesticides are challenging due to their similar chemistries and similar masses and mass fragments. For example, Thermo Scientific Dionex™ IonPac™ CS21-Fast-4μm cation-exchange column was designed to resolve diquat and paraquat. This technical note provides the IC-MS conditions for twelve of the fifteen analytes. Three analytes, 1,2,4-triazole, difenzoquat, and propamocarb, are unsuitable using the Dionex IonPac CS21-Fast-4μm column for IC-MS analysis.| Market: | Food and Beverage |
| Keywords: | Diethanolamine, Diquat, melamine, Mepiquat, TEA, Triazole, Quaternary amine, RFIC, DEA, IC-MS, Paraquat, triethanolamine, Cyromazine, Reagent-Free IC, polar pesticides, trimethylsulfonium, chlormequat, IonPac CS21, AN 000607, cationic polar pesticides, AN 001166, AN000607, TN 001521, TN 001900, TN001900, dimethylhydrazine, propamocarb, TMS, cationic polar pesticide, difenzoquat, IonPac CS21-Fast-4μm, nereistoxin, quaternary amine pesticide, AN001166, TN001521 |
| Matrix: | Standard (DI water diluent) |
| Author: | Terri Christison, John E. Madden, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.