Find methods for your needs
Improved analysis of quaternary amines using a Thermo Scientific BioBasic SCX HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of quaternary amines. The separation is performed on a Thermo Scientific BioBasic SCX HPLC column, with UV detection at 290 nm.| Market: | Environmental |
| Keywords: | BioBasic SCX, Diquat, HPLC, Paraquat, Quaternary amines, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.