Find methods for your needs
Rapid and cost-effective determination of Class 3 residual solvents in pharmaceutical products by HS-GC with hydrogen as carrier gas Be the first to rate this application
rapid costeffective determination class 3 residual solvents pharmaceutical products by hsgc with hydrogen as carrier gas
Description
The aim of this study is to demonstrate the suitability of the Thermo Scientific™ TriPlus™ 500 static headspace autosampler in combination to the Thermo Scientific™ TRACE™ 1610 GC to run an optimized rapid method for the determination of Class 3 residual solvents in pharmaceutical products
Market: Other
Keywords: GC, Gas Chromatography, USP <467>, TriPlus 500 headspace autosampler, Class 3 residual solvents, TRACE 1610 gas chromatograph, valve and loop static headspace, flame ionization detector (FID), hydrogen carrier gas, mass spectrometry detector
Matrix: Pharmaceutical products
Author: Giulia Riccardino[[1]], Daniela Cavagnino[[1]], Carlo Ceciliato[[1]], and Daniel Kutscher[[2]]
Affiliation: [[1]]Thermo Fisher Scientific, Milan, Italy, [[2]]Thermo Fisher Scientific, Milan, Italy
Uploaded on 3/20/2023.