Find methods for your needs
Biodiesel quality assessment: characterization of residual methanol in finished biodiesel (B100) by headspace sampling according to EN 14110 standard
Description
The aim of this study is to demonstrate the suitability of the Thermo Scientific™ TRACE™ 1610 GC series combined with the Thermo Scientific™ TriPlus™ RSH SMART autosampler for characterization of residual methanol in biodiesel according to the EN 14110:2020 method.| Market: | Petrochem, Oil and Gas |
| Keywords: | FID, GC, Methanol, biodiesel, Automated sample preparation, Headspace, Flame Ionization Detection, Gas Chromatography, TriPlus RSH SMART, B100, HeSaver-H2Safer, EN 14110 |
| Matrix: | Biodiesel (B 100) |
| Author: | Giulia Riccardino [[1]], Daniela Cavagnino [[1]], and Daniel Kutscher[[2]] |
| Affiliation: | [[1]] Thermo Fisher Scientific, Milan, Italy, [[2]] Thermo Fisher Scientific, Bremen, DE |